Learn More
Cayman Chemical 4-oxo-2-Nonal Alkyn 5mg
SDP
Supplier:  Cayman Chemical 171045 MG
Encompass_Preferred
An alkyne-tagged electrophile that can be used as a probe to isolate and identify the reaction products of lipid peroxidation using click chemistry
Catalog No. 50-298-0340