Learn More
Abcam 4-P-PDOT, MT2 selective melatonin receptor antagonist, 50MG
Supplier:  Abcam AB14641950MG
Encompass_Preferred
MW 279.4 Da, Purity >99%. MT2 selective melatonin receptor antagonist (Ki values are 0.46 and 56 nM for MT2 and MT1 respectively). Counteracts melatonin-mediated ERK- and Nrf2-related antioxidant potential *in vivo.* Centrally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4634