Learn More
Cayman Chemical 4-palmItamIdo TEMPO 5mg
SDP
Supplier:  Cayman Chemical 259955 MG
Encompass_Preferred
A lipid-soluble spin label; incorporates into vesicles and cell membranes; has been used to study the entrapment of molecules within phosphatidylcholine vesicles
Catalog No. 50-293-3383