Learn More
Medchemexpress LLC 4-Penten-1-ol | 821-09-0 | 99.99% | 86.13 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW010639
Encompass_Preferred
4-Penten-1-ol is commonly used as a flavor ingredient in various products, including food, beverages, and perfumes. It serves as a starting material for the synthesis of organic compounds like pharmaceuticals and agrochemicals. This compound has also been investigated for its potential as a biobased solvent due to low toxicity and biodegradability, and for its antimicrobial properties against certain bacteria and fungi.
  • Used as a flavor ingredient
  • Starting material for organic synthesis
  • Potential biobased solvent
  • Antimicrobial properties
Catalog No. 50-003-66643