Learn More
Medchemexpress LLC 4-Phenoxybenzoic acid | 2215-77-2 | 99.8% | C13H10O3 | 500 G
SDP
Supplier:  Medchemexpress LLC HY78854500G
Encompass_Preferred
4-Phenoxybenzoic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It can block DNA binding of the human papillomavirus (HPV) E2 protein and is employed as an intermediate of Sitafloxacin, a fluoroquinolone antibiotic used in the treatment of Buruli ulcer.
  • Used as a biological material or organic compound for life science research
  • Blocks DNA binding of human papillomavirus (HPV) E2 protein
  • Employed as an intermediate of Sitafloxacin, a fluoroquinolone antibiotic used in the treatment of Buruli ulcer
Catalog No. 50-004-34505