Learn More
Medchemexpress LLC 4-Phenoxybenzylamine | 107622-80-0 | 98.45% | 199.25 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1856350MG
Encompass_Preferred
4-Phenoxybenzylamine inhibits the function of the NS3 protein by stabilizing an inactive conformation. This action targets a highly conserved novel binding site located at the interface between the protease and helicase domains of the Hepatitis C Virus (HCV) NS3 protein, representing a new class of direct acting antiviral agents.
  • Inhibits NS3 protein function
  • Stabilizes inactive conformation of NS3 protein
  • Targets highly conserved novel binding site
  • Acts as a direct acting antiviral agent
Catalog No. 50-003-87781