Learn More
Medchemexpress LLC 4-phenyl-1,2,4-triazoline-3,5-dione | 4233-33-4 | MFCD00003148 | 98.0% | 175.14 | C8H5N3O2 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW016748100G
Encompass_Preferred
PTAD (4-Phenyl-1,2,4-triazoline-3,5-dione) is a dieneophile reagent used in organic synthesis and biochemical research. It functions as an oxidizing reagent for thiols, enabling conversion to disulfides, and participates in Diels-Alder and other cycloaddition reactions. The product is supplied as an orange-red solid with high purity for laboratory use.
  • Acts as a dieneophile for cycloaddition reactions.
  • Oxidizes thiols to form disulfides.
  • High purity (98.0%) suitable for research applications.
  • Orange to red solid form for easy visual identification.
  • Store at 4°C and protect from light to maintain stability.
Catalog No. 50-003-01153