Learn More
Medchemexpress LLC 4-phenyl-2-butanone | 2550-26-7 | MFCD00008790 | 99.8% | 148.20 g/mol | C10H12O | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01561610MM/1ML
Encompass_Preferred
Benzylacetone (4-phenyl-2-butanone, CAS 2550-26-7) is an aromatic ketone used in research as a tyrosinase inhibitor and as an intermediate in flavor and fragrance chemistry. It is a colorless to light yellow liquid with molecular weight 148.20 g/mol and is supplied as a high-purity reagent for biochemical and analytical applications.
  • High purity (99.8%).
  • Tyrosinase inhibitory activity reported.
  • Colorless to light yellow liquid, compatible with common organic solvents.
  • Soluble in DMSO for prepared stock solutions.
  • Suitable for biochemical assays and analytical studies.
Catalog No. 50-003-00732