Learn More
Medchemexpress LLC 4-phenyl-2-pyrrolidinone | 1198-97-6 | MFCD01687226 | 99.9% | 161.20 g/mol | C10H11NO | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01785110G
Encompass_Preferred
4-Phenyl-2-pyrrolidinone is a small-molecule synthetic intermediate used in medicinal chemistry and chemical synthesis. It is employed as a precursor for preparing compounds with anticonvulsant and nootropic activity and as an intermediate in the synthesis of bromodomain-containing protein 4 (BRD4) bromodomain inhibitors. Analytical documentation commonly provided includes a certificate of analysis, 1H NMR, RP-HPLC, and mass spectrometry.
  • High purity suitable for research and synthesis (reported ~99.9%).
  • Used as a precursor and synthetic intermediate in drug discovery.
  • Characterized by COA, 1H NMR, RP-HPLC, and MS analytical methods.
  • Molecular formula C10H11NO; molecular weight 161.20 g/mol.
Catalog No. 50-003-01598