Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-phenyl-2-pyrrolidinone | 1198-97-6 | MFCD01687226 | 99.9% | 161.20 g/mol | C10H11NO | 50 G
SDP

Supplier:  Medchemexpress LLC HYW01785150G

Encompass_Preferred

4-Phenyl-2-pyrrolidinone is a chemical intermediate and drug-precursor used in organic synthesis. It is a white to off-white solid and serves as a building block for the synthesis of anticonvulsant, nootropic, and bromodomain inhibitor scaffolds.

  • Molecular formula: C10H11NO.
  • Molecular weight: 161.20 g/mol.
  • Appearance: white to off-white solid.
  • Purity: 99.93%.
  • Applications: precursor and intermediate in medicinal chemistry and organic synthesis.
  • Structural representation: SMILES O=C1NCC(C2=CC=CC=C2)C1.

Catalog No. 50-003-00617


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.