Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-phenyl-7,8-dihydroxycoumarin | 842-01-3 | MFCD02058070 | 99.4% | 254.24 g·mol⁻¹ | C15H10O4 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1284105MG

Encompass_Preferred

4-phenyl-7,8-dihydroxycoumarin is a coumarin derivative provided for research use. It is used in bronchiectasis-related studies and supplied as an off-white to yellow solid or as a DMSO solution, with high purity and defined storage recommendations.

  • High purity: 99.4%.
  • Molecular formula: C15H10O4; molecular weight: 254.24 g·mol⁻¹.
  • Physical form: off-white to yellow solid; also available as 10 mM solution in DMSO.
  • Typical pack sizes: 5 mg, 10 mg, 50 mg, 100 mg.
  • Storage: powder at -20°C (stable up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Intended use: for laboratory research only.

Catalog No. 50-002-64111


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.