Learn More
Medchemexpress LLC 4-(phenylamino)phenol | 122-37-2 | MFCD00020142 | 98.0% | 185.22 g/mol | C12H11NO | 5 G

Supplier: Medchemexpress LLC HYW0883205G
4-Hydroxydiphenylamine (p-anilinophenol) is an aromatic amine supplied as a solid reagent for research and laboratory use. It functions as an antioxidant and as an intermediate or catalyst in organic synthesis, and it is used in environmental science and toxicology studies.
- Purity: 98.02% (analytical)
- CAS number: 122-37-2
- Chemical formula: C12H11NO
- Molecular weight: 185.22 g/mol
- Physical form: solid (powder)
- Typical quantity: 5 g pack
- Typical applications: antioxidant research, chemical synthesis intermediate, environmental and toxicology studies
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.