Learn More
Medchemexpress LLC 4-Phenylbutyric acid | 1821-12-1 | 99.98% | C10H12O2 | 10MM/1ML
SDP
Supplier:  Medchemexpress LLC HYA028110MM/1ML
Encompass_Preferred
4-Phenylbutyric acid (4-PBA) is an inhibitor of histone deacetylase (HDAC) and endoplasmic reticulum (ER) stress. It is used in cancer and infection research.
  • Inhibits histone deacetylase (HDAC)
  • Inhibits endoplasmic reticulum (ER) stress
  • Used for cancer research
  • Used for infection research
  • Suitable for laboratory chemical applications
Catalog No. 50-004-33447