Learn More
Medchemexpress LLC HY-A0281 500mg , 4-Phenylbutyric acid CAS:1821-12-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYA0281500MG
Encompass_Preferred
HY-A0281 500mg 4-Phenylbutyric acid CAS: 1821-12-1 4-Phenylbutyric acid (4-PBA) is an inhibitor of HDAC and endoplasmic reticulum (ER) stress, used in cancer and infection research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0279