Learn More
Medchemexpress LLC 4- Phenyldiazenyl be 10mM 1mL
SDP
Supplier:  Medchemexpress LLC HYW10623410MM/1ML
Encompass_Preferred
4-(Phenyldiazenyl)benzoic acid is a photosensitive and photoswitchable TRPA1 agonist that can be used as pharmacological tools for study of pain signaling
Catalog No. 50-002-06952