Learn More
Cole-Parmer Velp F105A0108 JLT4 Laboratory 4-Position Flocculator 100-240V
Supplier:  Cole-Parmer 9956113
Encompass
Jar testers are designed to test the efficiency of flocculation or precipitation reagents on a small scale. Stirring rod shafts can individually be stopped and height can be adjusted while the system is operating. Choose from portable or laboratory testers.
Catalog No. NC1784813