Learn More
JDM LAB SOLUTIONS LLC 4 POSITION-MULTI MAG STIRRER
Supplier:  JDM LAB SOLUTIONS LLC MS4BSUA
Encompass
4 Position Magnetic Stirrer. The front control panel has the ability to detach from the main unit. This unique feature allows the main stirring base to be placed & operated in a water bath.
Catalog No. NC1866749