Learn More
Medchemexpress LLC 4-propylphenylboronic acid | 134150-01-9 | MFCD00859261 | 99.9% | 164.01 | C9H13BO2 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01538010G
Encompass_Preferred
4-Propylphenylboronic acid is an aromatic boronic acid supplied as a high-purity biochemical reagent for research use. It functions as a synthetic building block and is commonly used in boron-mediated coupling reactions and assay development. Not for human or clinical use.
  • High purity (99.9%) suitable for research applications.
  • Molecular formula C9H13BO2; molecular weight 164.01.
  • Supplied as a crystalline white to off-white powder.
  • Used as a building block in Suzuki-Miyaura coupling and other organic syntheses.
  • Datasheet, certificate of analysis, and safety data sheet available.
Catalog No. 50-003-03268