Learn More
Medchemexpress LLC 4-Propylphenylboronic acid | 134150-01-9 | MFCD00859261 | 99.9% | 164.01 g·mol⁻¹ | C9H13BO2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW015380
Encompass_Preferred
4-Propylphenylboronic acid is an organoboronic acid used as a building block and reagent in organic synthesis and life-science research. Supplied as a high-purity crystalline solid, it is commonly applied in Suzuki-Miyaura cross-coupling and related transformations. Manufacturer documentation includes analytical data and handling instructions for research use.
  • High purity: 99.9%.
  • Suitable for Suzuki-Miyaura cross-coupling and other C-C bond-forming reactions.
  • Supplied as a crystalline solid for easy weighing and storage.
  • Provided with analytical data (H-NMR, RP-HPLC) and handling guidance.
  • Intended for research use only; not for human or diagnostic use.
Catalog No. 50-003-04117