Learn More
Medchemexpress LLC 4-Pyridinecarboxylic acid, 2-[3-[2-chloro-4-[[5-cyclopropyl-3-(2,6-dichlorophenyl)-4-isoxazolyl]methoxy]phenyl]-3-hydroxy-1-azetidinyl | 1418274-28-8 | MFCD31731099 | 99.8% | 586.85 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY109083
Encompass_Preferred
Cilofexor is a potent, selective, and orally active nonsteroidal FXR agonist with an EC50 of 43 nM. It exhibits anti-inflammatory and antifibrotic effects and has potential for research in primary sclerosing cholangitis (PSC) and nonalcoholic steatohepatitis (NASH).
  • Potent, selective, and orally active nonsteroidal FXR agonist
  • Exhibits anti-inflammatory and antifibrotic effects
  • Potential for research in primary sclerosing cholangitis
  • Potential for research in nonalcoholic steatohepatitis
  • Involved in various clinical trials
Catalog No. 50-003-47555