Learn More
Medchemexpress LLC 4-Pyridinemethanamine, N-[(3S)-1-[2-(5-chloro-2,4-dimethoxyphenyl)imidazo[1,2-a]pyridin-7-yl]-3-pyrrolidinyl] | 99.6% | 522.31 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY122181B
Encompass_Preferred
OTS186935 hydrochloride is a potent protein methyltransferase SUV39H2 inhibitor with an IC50 of 6.49 nM. It significantly inhibits tumor growth in mouse xenograft models without detectable toxicity and regulates the production of γ-H2AX in cancer cells.
  • Potent protein methyltransferase SUV39H2 inhibitor (IC50 of 6.49 nM).
  • Shows significant inhibition of tumor growth in mouse xenograft models without detectable toxicity.
  • Regulates the production of γ-H2AX in cancer cells.
Catalog No. 50-004-39184