Learn More
Medchemexpress LLC 4-pyrimidinamine, 6-chloro-N-(phenylmethyl)- | 61667-16-1 | MFCD00679879 | 99.7% | 219.67 g/mol | C11H10ClN3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW02759550MG
Encompass_Preferred
N-Benzyl-6-chloropyrimidin-4-amine is a pyrimidine-derived research compound used as a building block in medicinal chemistry and biochemical screening. It is supplied as a solid or as a DMSO solution and features a chlorinated pyrimidine core.
  • High reported purity suitable for research use.
  • Available in small sample and mg-scale quantities.
  • Off-white solid appearance for easy handling.
  • Provided as a 10 mM solution in DMSO for screening workflows.
  • Suitable as a synthetic intermediate or screening reagent.
  • Certificate of analysis available for batch traceability.
Catalog No. 50-003-00138