Learn More
Medchemexpress LLC 4-pyrimidine methanamine | 45588-79-2 | MFCD06796202 | 99.3% | 109.13 g·mol⁻¹ | C5H7N3 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW017769500MG
Encompass_Preferred
4-Pyrimidine methanamine (pyrimidin-4-ylmethanamine) is a small-molecule synthetic intermediate used in pharmaceutical research and chemical synthesis. Supplied for research use only, it is intended as an intermediate for laboratory and medicinal chemistry applications.
  • High purity (99.3%).
  • Chemical formula C5H7N3; molecular weight 109.13 g·mol⁻1.
  • Light yellow to brown liquid appearance.
  • Suitable as a building block in medicinal chemistry and synthetic workflows.
  • Available in small-scale pack sizes for laboratory use.
  • Not for human or clinical use; research use only.
Catalog No. 50-003-01668