Learn More
Medchemexpress LLC 4-quinazolinamine, 6-(2-chloroethoxy)-N-(3-ethynylphenyl)-7-(2-methoxyethoxy) | 183321-83-7 | 98.7% | 10 MG

Supplier: Medchemexpress LLC HY12008Y10MG
Erlotinib impurity 45 is an analytical reference impurity of erlotinib supplied as a solid reference standard for research use only. It is intended for impurity characterization, method development, and quality-control studies.
- Analytical reference for erlotinib impurity characterization.
- High purity: 98.7% (LCMS).
- Molecular formula C21H20ClN3O3, molecular weight 397.85.
- Available in milligram pack sizes, for example 10 MG.
- Recommended storage: 4°C under nitrogen; in solvent, store at -80°C for up to 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.