Learn More
Medchemexpress LLC 4-quinolinecarboxamide, 2-(4-ethoxyphenyl)-n-[3-(2-methyl-1-piperidinyl)propyl]- | 689270-18-6 | 99.9% | 431.57 g/mol | C27H33N3O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY14965210MG
Encompass_Preferred
IRF5-IN-1 is a small-molecule research compound that blocks IRF5 activation and downstream TLR7/8 signaling, producing anti-inflammatory effects observed in in vitro studies. Supplied as an off-white to light yellow solid, it is intended for biochemical and cell-based assays after dissolution in appropriate solvents (DMSO recommended). Follow recommended storage and handling conditions for experimental reproducibility.
  • Selective inhibition of IRF5-mediated signaling in cellular assays.
  • High purity (~99.9%) suitable for research applications.
  • Soluble in DMSO at 50 mg/mL with ultrasonic warming to 80°C.
  • Supplied as a 10 mg powder for small-scale experiments.
  • Stable when stored as powder at -20°C, with defined in-solution stability.
  • Useful tool for studying TLR7/8-IRF5 inflammatory pathways.
Catalog No. 50-002-86061