Learn More
Medchemexpress LLC 4-tert-Butyl-benzhyd 10mM 1mL | 62034-73-5 | 193.24 g/mol | C11H15NO2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11481810MM 1ML
Encompass_Preferred
4-(tert-Butyl)-benzhydroxamic acid is a benzhydroxamic acid derivative supplied as a ready-to-use 10 mM solution in DMSO for research use. It is intended for biochemical and cellular assays where hydroxamic acid moieties act as metal-binding groups or modulators of enzyme activity.
  • Provided as a 10 mM solution in DMSO (1 mL).
  • Also available as solid powder in multiple milligram pack sizes (5, 10, 50, 100 mg).
  • High purity suitable for research applications (reported 99.49%).
  • CAS number 62034-73-5 for substance identification.
Catalog No. 50-002-29397