Learn More
Medchemexpress LLC 4-tert-Butyl-benzhyd 50mg | 62034-73-5 | 193.24 g/mol | C11H15NO2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11481850MG
Encompass_Preferred
4-(tert-Butyl)-benzhydroxamic acid is a hydroxamic acid derivative used as a research reagent. It has been reported to act as a PqsR antagonist that modulates bacterial quorum sensing and reduces pyocyanin production in Pseudomonas aeruginosa. Supplied as a solid or as solution aliquots, it is intended for in vitro research use only.
  • Reported PqsR antagonist activity with micromolar IC50s in bacterial assays.
  • High purity (99.49%) suitable for research applications.
  • Available as neat solid and 10 mM solutions in DMSO for screening and assays.
  • Multiple pack sizes available for flexibility in experiments (for example, 5 mg, 10 mg, 50 mg).
  • Chemical formula C11H15NO2; molecular weight 193.24 g/mol.
Catalog No. 50-002-26407