Learn More
Medchemexpress LLC 4-tert-Butyl-benzhyd 5mg | 62034-73-5 | 5 MG

Supplier: Medchemexpress LLC HY1148185MG
4-(tert-butyl)-benzhydroxamic acid is a benzhydroxamic acid derivative used in bacterial quorum sensing research as a PqsR antagonist. It is supplied as a solid research reagent with high purity and is commonly used to study pyocyanin production and related virulence pathways.
- High purity: 99.5%
- Molecular weight: 193.24 g/mol
- Chemical formula: C11H15NO2
- CAS number: 62034-73-5
- Available sizes: solid quantities from 5 mg to 500 mg
- Documented resources: datasheet, certificate of analysis, and safety data sheet available
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.