Learn More
Medchemexpress LLC 4-tert-butylcalix[4]arene-tetraacetic acid tetraethyl ester | 97600-39-0 | MFCD00145373 | >=97.0% | 993.3 g/mol | C60H80O12 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW020249
Encompass_Preferred
Sodium ionophore X is a calixarene-derived sodium-selective ion carrier used in analytical chemistry and membrane transport research. It is commonly supplied as the tetraethyl ester of 4-tert-butylcalix[4]arene-tetraacetic acid and is employed in ion-selective electrodes and related membrane assays for sodium detection and transport studies.
  • Sodium-selective ion carrier suitable for ion-selective electrodes and membrane studies.
  • Reported purity of ≥97.0% (HPLC) in supplier descriptions.
  • Molecular formula C60H80O12 and molecular weight 993.3 g/mol.
  • Typically provided as the tetraethyl ester derivative for membrane incorporation.
Catalog No. 50-003-03647