Learn More
Medchemexpress LLC 4-tert-Butylphenylboronic acid | 123324-71-0 | 178.04 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00738910MM/1ML
Encompass_Preferred
4-tert-Butylphenylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.
  • Biochemical reagent
  • Can be used as a biological material or organic compound
  • Applicable for life science related research
  • Powder storage at -20°C for 3 years or 4°C for 2 years
  • In solvent storage at -80°C for 6 months or -20°C for 1 month
  • Stock solution storage at -80°C for 6 months or -20°C for 1 month, with aliquoting recommended
Catalog No. 50-004-60342