Learn More
Medchemexpress LLC 4-tert-Butylphenylboronic acid | 123324-71-0 | 178.04 | 100 G

Supplier: Medchemexpress LLC HYW007389100G
4-tert-Butylphenylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. This white to off-white solid has a molecular weight of 178.04 and a purity of 99.96%.
- Used as a biochemical reagent
- Applicable as a biological material or organic compound
- Suitable for life science research
- For research use only
- Not for patient use
- Molecular weight: 178.04
- Formula: C10H15BO2
- Appearance: White to off-white solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.