Learn More
Medchemexpress LLC 4-tert-Butylphenylboronic acid | 123324-71-0 | 178.04 | 25 G

Supplier: Medchemexpress LLC 25GHYW007389
4-tert-Butylphenylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is intended for research use only.
- Used as a biochemical reagent
- Can be used as a biological material or organic compound
- Intended for life science related research
- For research use only
- Storage (powder): -20°C for 3 years, 4°C for 2 years
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.