Learn More
Medchemexpress LLC 4-tert-Butylphenylboronic acid | 123324-71-0 | 178.04 | 5 G

Supplier: Medchemexpress LLC HYW0073895G
4-tert-Butylphenylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
- Appears as a solid
- Color is white to off-white
- Recommended storage for powder is -20°C for 3 years, or 4°C for 2 years
- Recommended storage for in solvent is -80°C for 6 months, or -20°C for 1 month
- Soluble in DMSO at 25 mg/mL (140.42 mM), requiring ultrasonic, warming, and heating to 60°C
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.