Learn More
Medchemexpress LLC 4-tert-Octylphenol | 140-66-9 | 99.7% | 1000 G
SDP
Supplier:  Medchemexpress LLC HYB19411000G
Encompass_Preferred
4-tert-Octylphenol is an endocrine-disrupting chemical and an estrogenic agent, also a biodegradation product of non-ionic surfactants alkylphenol polyethoxylates. It induces apoptosis in neuronal progenitor cells in offspring mouse brain, reduces bromodeoxyuridine (BrdU), mitotic marker Ki67, and phospho-histone H3 (p-Histone-H3), which leads to a reduction of neuronal progenitor proliferation. 4-tert-Octylphenol disrupts brain development and behavior in mice.
  • Endocrine-disrupting chemical
  • Estrogenic agent
  • Biodegradation product of non-ionic surfactants alkylphenol polyethoxylates
  • Induces apoptosis in neuronal progenitor cells in offspring mouse brain
  • Reduces neuronal progenitor proliferation by affecting BrdU, Ki67, and phospho-histone H3
  • Disrupts brain development and behavior in mice
  • Promising for research of immune response, neuro-related diseases and ethology
Catalog No. 50-004-11783