Learn More
Medchemexpress LLC 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[[3-(2,6-dichlorophenyl)-5-methyl-4-isoxazolyl]carbonyl]amino]-3,3-dimethyl-7-oxo-, sodium salt (1:1), (2S,5R,6R)- | 343-55-5 | 98.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB14595MG
Encompass_Preferred
Dicloxacillin sodium is a β-lactam antibiotic belonging to the penicillin family. It is effective against Gram-positive bacteria, including β-lactamase-producing organisms such as *Staphylococcus aureus*.
  • β-lactam antibiotic.
  • Active against Gram-positive bacteria.
  • Effective against β-lactamase-producing organisms like *Staphylococcus aureus*.
Catalog No. 50-004-13355