Learn More
Medchemexpress LLC 4-Thiazolecarboxamide, 5-(3-methoxypropyl)-2-phenyl-N-[2-[6-(1-pyrrolidinylmethyl)thiazolo... | 2070015-26-6 | 99.85% | 606.20 | 50 MG

Supplier: Medchemexpress LLC HY19758A50MG
Sirtuin modulator 1 acts as a modulator of SIRT1, a homolog of SIRT3, with an EC1.5 of less than 1 μM. This compound is derived from patent WO 2010071853 A1, specifically Compound No.4.
- Modulates SIRT1 with high potency
- Homolog to SIRT3
- Appears as a light yellow to yellow solid
- Can be stored at 4°C, sealed, and away from moisture
- Recommended storage in solvent at -80°C for 6 months or -20°C for 1 month
- Soluble in DMSO for in vitro applications
- Demonstrates solubility in various in vivo protocols
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.