Learn More
Medchemexpress LLC 4-thiazolecarboxamide, N-(2-aminoethyl)-5-(3-fluorophenyl)-, hydrochloride | 127917-66-2 | MFCD00900596 | 100.0% | 301.77 | C12H13ClFN3OS | 10 MG
SDP
Supplier:  Medchemexpress LLC HY100027A10MG
Encompass_Preferred
Ro 41-1049 hydrochloride is the hydrochloride salt of a reversible, selective monoamine oxidase A (MAO-A) inhibitor supplied as a white to off-white solid for research use. The compound is intended for biochemical and pharmacological studies, including receptor binding and enzyme activity assays, and is provided with supporting analytical documentation.
Catalog No. 50-002-15134