Learn More
Medchemexpress LLC 4-Thiouracil | 591-28-6 | 99.98% | 128.15 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW010417
Encompass_Preferred
4-Thiouracil is a thionucleobase with cytostatic properties. It can be used as biological photoprobes to detect RNA structures and nucleic acid-nucleic acid contacts. It also acts as a strong ultraviolet A (UVA) photosensitizer, providing a source of the reactive oxygen species of O2. It is promising for research in photocrosslinking, photodamage, and photodynamic therapy.
  • Can be used as biological photoprobes to detect RNA structures and nucleic acid-nucleic acid contacts.
  • Acts as a strong ultraviolet A (UVA) photosensitizer, providing a source of reactive oxygen species (O2).
  • Promising for research in photocrosslinking, photodamage, and photodynamic therapy.
Catalog No. 50-003-66658