Learn More
Medchemexpress LLC 4-Thiouridine | 13957-31-8 | 99.9% | 260.27 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW011793100MG
Encompass_Preferred
4-Thiouridine is a ribonucleoside analog widely utilized in RNA analysis and (m)RNA labeling. It is known to inhibit rRNA synthesis and induce a nucleolar stress response.
  • Widely used in RNA analysis and (m)RNA labeling.
  • Inhibits rRNA synthesis and causes a nucleolar stress response.
Catalog No. 50-004-53815