Learn More
Medchemexpress LLC 4-Thiouridine | 13957-31-8 | 100.0% | 260.27 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW0117935MG
Encompass_Preferred
4-Thiouridine is a ribonucleoside analog widely used in RNA analysis and (m)RNA labeling. It inhibits rRNA synthesis and causes a nucleolar stress response.
  • Ribonucleoside analog
  • Used in RNA analysis and (m)RNA labeling
  • Inhibits rRNA synthesis
  • Causes a nucleolar stress response
  • At 0-10 μM, it labels and purifies nascent rRNA without significantly affecting rRNA synthesis
  • At ≥50 μM, it strongly inhibits production and processing of rRNA
Catalog No. 50-004-52610