Learn More
Medchemexpress LLC 4-(trifluoromethoxy)aniline | 461-82-5 | MFCD00041314 | >98.0% | 177.13 g·mol⁻¹ | C7H6F3NO | 500 G
SDP
Supplier:  Medchemexpress LLC HYW016664500G
Encompass_Preferred
4-(Trifluoromethoxy)aniline is an aromatic amine used as an intermediate and analytical standard in chemical research. It introduces a para-trifluoromethoxy group into aniline scaffolds and is employed in medicinal chemistry and agrochemical synthesis. The compound is supplied in high-purity grades for laboratory synthesis and analytical applications.
  • Aromatic aniline derivative with a para-trifluoromethoxy substituent.
  • High purity suitable for synthesis and analytical use.
  • Liquid appearance, straightforward handling under standard laboratory conditions.
  • Useful building block for medicinal chemistry and agrochemical research.
  • Compatible with common synthetic transformations for small-molecule construction.
Catalog No. 50-003-03237