Learn More
Medchemexpress LLC 4-(Trifluoromethyl)aniline-d4 | 1219795-48-8 | 99.23% | C7H2D4F3N | 25 MG

Supplier: Medchemexpress LLC 25MGHY77324S
4-(Trifluoromethyl)aniline-d4 is a deuterium-labeled compound, also known as 4-Aminobenzotrifluoride-d4. It is a DYRK1A and DYRK1B kinase inhibitor with IC50 values of 54.84 nM and 186.40 nM, respectively. This compound can serve as a tracer or internal standard for quantitative analysis using methods such as NMR, GC-MS, or LC-MS.
- Deuterium-labeled compound
- DYRK1A and DYRK1B kinase inhibitor
- Purity of 99.23%
- Suitable for use as a tracer or internal standard
- Ideal for quantitative analysis via NMR, GC-MS, or LC-MS
- Store pure form at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.