Learn More
Medchemexpress LLC 4-(trifluoromethyl)aniline-d4 | 1219795-48-8 | MFCD03428115 | 99.2% | 165.15 g/mol | C7H6F3N | 10 MG
SDP
Supplier:  Medchemexpress LLC HY77324S10MG
Encompass_Preferred
4-(Trifluoromethyl)aniline-d4 is a deuterium-labeled (d4) derivative of 4-(trifluoromethyl)aniline supplied for laboratory research use as an isotopic/internal standard and research reagent. It is intended for analytical applications such as mass spectrometry and synthetic chemistry; not for human or clinical use.
  • Deuterium-labeled (d4) isotopic analog for isotope dilution and tracking.
  • High chemical purity: 99.2% (GC) per supplier certificate of analysis.
  • Useful as an internal standard for mass spectrometry and analytical quantitation.
  • Provided in a 10 MG vial for laboratory use.
Catalog No. 50-002-99274