Learn More
Medchemexpress LLC 4-(trifluoromethyl)benzoic acid-13C6 | 2731552-41-1 | MFCD32201889 | 98.3% | 196.08 | C213C6H5F3O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW015198S
Encompass_Preferred
4-(Trifluoromethyl)benzoic acid-13C6 is the 13C6 isotopically labeled form of 4-(trifluoromethyl)benzoic acid. It is intended for use as a stable-isotope internal standard and tracer in analytical and synthetic chemistry, providing a predictable mass shift for accurate quantitation and method development. The product is supplied with analytical data and handling documentation to support laboratory use.
  • Stable 13C6 isotopic labeling for accurate mass spectrometry quantitation.
  • High purity supporting reliable analytical performance.
  • Available in small quantities suitable for standards and method development.
  • Includes analytical data and documentation (COA, HNMR, HPLC, MS, SDS).
  • Suitable as an internal standard, tracer, and reference material.
Catalog No. 50-003-02905