Learn More
Medchemexpress LLC 4-(trifluoromethyl)-L-phenylalanine | 114926-38-4 | MFCD00044946 | 99.9% | 233.19 | C10H10F3NO2 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0139621G
Encompass_Preferred
4-(Trifluoromethyl)-L-phenylalanine is a fluorinated derivative of the amino acid L-phenylalanine used as a research reagent and building block for peptide synthesis and chemical biology studies. It has molecular formula C10H10F3NO2, molecular weight 233.19, and CAS 114926-38-4.
  • Fluorinated amino acid for incorporation into peptides and small molecules.
  • High purity (99.89%) suitable for analytical and synthetic applications.
  • White to off-white solid with defined appearance for handling consistency.
  • Available in multiple pack sizes to support varied research scales.
  • Useful for modulating hydrophobicity and electronic properties in peptide design.
Catalog No. 50-003-03937