Learn More
Medchemexpress LLC 4-(trifluoromethyl)-L-phenylalanine | 114926-38-4 | MFCD00044946 | 99.9% | 233.19 g·mol⁻1 | C10H10F3NO2 | 500 MG

Supplier: Medchemexpress LLC HYW013962500MG
4-(Trifluoromethyl)-L-phenylalanine is an L-phenylalanine derivative bearing a para-trifluoromethyl substituent on the aromatic ring. Provided as a high-purity research reagent, it is intended for use in peptide synthesis, chemical biology studies, and experiments requiring modified amino acids.
- High purity suitable for research applications.
- Molecular weight 233.19 g·mol⁻1.
- Chemical formula C10H10F3NO2.
- Available in multiple sizes for scalability of experiments.
- Useful as an amino acid analog in peptide synthesis and biochemical studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.