Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific 4-(Trifluoromethyl)phenylhydrazine, 368-90-1, MFCD00042508, 5g

Supplier:  Matrix Scientific 0054595G

Encompass_Preferred

Molecular Formula C7H7F3N2, Purity 97%, Molecular Weight 176.14, Melting Point 63-65°, Boiling Point 118-122/17mm

Catalog No. 50-011-8076


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.