Learn More
Medchemexpress LLC 4-(Trifluoromethyl)umbelliferone | 575-03-1 | 98.7% | 5 G

Supplier: Medchemexpress LLC HYD01495G
4-(Trifluoromethyl)umbelliferone is a fluorescent probe substrate specifically used for rat hepatic cytochrome P450 enzymes. It is intended for research use only and not for patient use.
- Fluorescent probe substrate for rat hepatic cytochrome P450 enzymes.
- Appearance: Solid
- Color: Pale purple to light pink
- Emission (Em): 460
- Excitation (Ex): 335
- Soluble in DMSO (≥ 100 mg/mL).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.