Learn More
Medchemexpress LLC 4-trifluoromethylsalicylic acid | 328-90-5 | MFCD01529657 | 99.9% | 206.12 g/mol | C8H5F3O3 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW014632
Encompass_Preferred
4-Trifluoromethylsalicylic acid (desacetyl triflusal) is a research-grade small molecule described as a platelet aggregation inhibitor. Supplied as a solid, it has the molecular formula C8H5F3O3 and a molecular weight of 206.12 g/mol. Intended for laboratory research use only, the material is provided with a high assay purity and recommended storage to maintain stability.
  • White to off-white solid suitable for analytical and biological research.
  • Molecular formula C8H5F3O3; molecular weight 206.12 g/mol.
  • High purity (99.93%) for reproducible experimental results.
  • Stable when stored as powder at 4°C for 2 years or -20°C for 3 years.
  • Used as a platelet aggregation inhibitor and as a metabolite/reference in mechanistic studies.
Catalog No. 50-003-02347