Learn More
Nanocomposix BioReady Gold Nanospheres - Streptavidin - 40 nm, 10 OD in aqueous conjugation buffer, 200 µL
SDP
Supplier:  Nanocomposix AUIR40200U
Encompass_Preferred
As a global leader in gold nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.40 nm streptavidin-conjugated spherical gold nanoparticles combine strong plasmonic optical properties with readily biofunctionalized surface chemistry. These nanoparticles are used in applications such as immunodiagnostics, lateral flow assays, biosensors, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, & UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.
Catalog No. NC1297357